Learn More
Chemglass Life Sciences 0.5 mL Cryogenic Vial, Self-Standing, External Thread, Sterile, Permanent 2D Barcode (Racked)
SDP
Catalog No. 502733388
Encompass_Preferred
This 0.5 mL sterile cryogenic vial is designed for reliable low-temperature storage and automated sample identification. The self-standing polypropylene vial features a permanent 2D barcode on the bottom for scanning while in racks, and an external-thread cap engineered without a silicone ring to reduce contamination risk. Vials are autoclavable and free of DNase, RNase, and human DNA.
  • Capacity 0.5 mL.
  • Permanent 2D barcode on vial bottom for in-rack scanning.
  • Sterile (E-beam sterilized, SAL 10^-6); non-pyrogenic; DNase/RNase and human DNA free.
  • USP VI polypropylene construction, autoclavable.
  • Self-standing, round bottom design for stability.
  • External thread cap without silicone ring to minimize contamination.
  • 9 x 9 vials per rack, 16 racks per case (1,296 vials per case).
  • Solid one-piece construction protects barcode during low-temperature storage.
Catalog No. 50-273-3388 Supplier Chemglass Life Sciences Supplier No. CGN476301
This 0.5 mL sterile cryogenic vial is designed for reliable low-temperature storage and automated sample identification. The self-standing polypropylene vial features a permanent 2D barcode on the bottom for scanning while in racks, and an external-thread cap engineered without a silicone ring to reduce contamination risk. Vials are autoclavable and free of DNase, RNase, and human DNA.
  • Capacity 0.5 mL.
  • Permanent 2D barcode on vial bottom for in-rack scanning.
  • Sterile (E-beam sterilized, SAL 10^-6); non-pyrogenic; DNase/RNase and human DNA free.
  • USP VI polypropylene construction, autoclavable.
  • Self-standing, round bottom design for stability.
  • External thread cap without silicone ring to minimize contamination.
  • 9 x 9 vials per rack, 16 racks per case (1,296 vials per case).
  • Solid one-piece construction protects barcode during low-temperature storage.