Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 0.5 mL, Self-Standing, Internal Thread, Sterile, 2D Barcode (CGN-4763-22)
SDP
Catalog No. 502733368
Encompass_Preferred
Chemglass Life Sciences CGN-4763-22 is a sterile 0.5 mL cryogenic vial engineered for reliable low-temperature sample storage and automated sample tracking. The self-standing vial features an internal thread and a one-piece cap design, and each vial is permanently laser-marked with a unique 2D barcode. Manufactured from USP VI polypropylene, the vial is autoclavable and certified free of DNase, RNase, and human DNA to reduce contamination risk.
  • 0.5 mL capacity for small-volume samples.
  • Permanently laser-marked 2D barcode for unique identification.
  • Self-standing design for stable handling and automated workflows.
  • Internal thread with one-piece cap to minimize contamination risk.
  • USP VI polypropylene construction, autoclavable.
  • Sterile (E-beam, SAL 10⁻⁶) and free of DNase, RNase, and human DNA.
  • Bulk-packed nests for efficient storage and handling (50/pk, 500/box, 2000/case).
Catalog No. 50-273-3368 Supplier Chemglass Life Sciences Supplier No. CGN476322
Chemglass Life Sciences CGN-4763-22 is a sterile 0.5 mL cryogenic vial engineered for reliable low-temperature sample storage and automated sample tracking. The self-standing vial features an internal thread and a one-piece cap design, and each vial is permanently laser-marked with a unique 2D barcode. Manufactured from USP VI polypropylene, the vial is autoclavable and certified free of DNase, RNase, and human DNA to reduce contamination risk.
  • 0.5 mL capacity for small-volume samples.
  • Permanently laser-marked 2D barcode for unique identification.
  • Self-standing design for stable handling and automated workflows.
  • Internal thread with one-piece cap to minimize contamination risk.
  • USP VI polypropylene construction, autoclavable.
  • Sterile (E-beam, SAL 10⁻⁶) and free of DNase, RNase, and human DNA.
  • Bulk-packed nests for efficient storage and handling (50/pk, 500/box, 2000/case).