Learn More
Bellco Glass, Inc. 2D Barcode 0.5ml Cryogenic Vial SelfStanding Internal Thread Sterile 9*9/rack 16 racks /cs 1296 vials/cs
SDP
Catalog No. 502329426
Encompass_Preferred
2D Barcode 0.5ml Cryogenic Vial SelfStanding Internal Thread Sterile 9*9/rack 16 racks /cs 1296 vials/cs
Catalog No. 50-232-9426 Supplier Bellco Glass, Inc. Supplier No. 451400536
2D Barcode 0.5ml Cryogenic Vial SelfStanding Internal Thread Sterile 9*9/rack 16 racks /cs 1296 vials/cs