Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 1.2 mL, Self-Standing, Internal Thread, Sterile, Laser 2D Barcode, Racked
SDP
Catalog No. 502733379
Encompass_Preferred
1.2 mL sterile cryogenic vial designed for secure long-term and ultra-low temperature sample storage. The self-standing, internally threaded vial features a silicone O-ring screw cap and a permanent laser-etched 2D barcode for reliable inventory tracking; vials are supplied racked for scanning without removal.
  • 1.2 mL nominal volume.
  • Self-standing, round bottom design for stable placement.
  • Internal thread with screw cap and silicone O-ring for a secure seal.
  • Permanent laser-etched 2D barcode on the vial bottom; scannable in rack.
  • USP VI polypropylene construction; non-pyrogenic and DNase/RNase and human DNA free.
  • Sterilized by E-beam (SAL=10^-6) and autoclavable.
  • Clear graduations and white marking area for sample labeling.
  • Packaged 100 vials per rack (10 x 10), 14 racks per case; 1,400 vials per case.
Catalog No. 50-273-3379 Supplier Chemglass Life Sciences Supplier No. CGN476310
1.2 mL sterile cryogenic vial designed for secure long-term and ultra-low temperature sample storage. The self-standing, internally threaded vial features a silicone O-ring screw cap and a permanent laser-etched 2D barcode for reliable inventory tracking; vials are supplied racked for scanning without removal.
  • 1.2 mL nominal volume.
  • Self-standing, round bottom design for stable placement.
  • Internal thread with screw cap and silicone O-ring for a secure seal.
  • Permanent laser-etched 2D barcode on the vial bottom; scannable in rack.
  • USP VI polypropylene construction; non-pyrogenic and DNase/RNase and human DNA free.
  • Sterilized by E-beam (SAL=10^-6) and autoclavable.
  • Clear graduations and white marking area for sample labeling.
  • Packaged 100 vials per rack (10 x 10), 14 racks per case; 1,400 vials per case.