Learn More
Chemglass Life Sciences Chemglass 1.2 mL Cryogenic Vial with 2D Barcode - Self-Standing, External Thread, Sterile (50/pack, 500/box)
SDP
Catalog No. 502733372
Encompass_Preferred
Chemglass CGN-4763-17 is a 1.2 mL cryogenic vial with a permanent laser-etched 2D barcode designed for reliable sample identification and handling. The vial is constructed from USP VI polypropylene and is sterilized by E-beam (SAL=10^-6). Packaged for laboratory use in multiple pack sizes to support inventory and storage workflows.
  • 1.2 mL capacity.
  • Laser-etched 2D barcode on the bottom for permanent identification.
  • USP VI polypropylene construction.
  • Sterile by E-beam; non-pyrogenic, DNase/RNase and human DNA free.
  • Self-standing round bottom with external thread.
  • Autoclavable.
  • Clear graduations and white marking area.
  • Special screw cap designed without a silicone ring to reduce contamination risk.
  • Packaged 50 per pack, 500 per box, 2000 per case.
Catalog No. 50-273-3372 Supplier Chemglass Life Sciences Supplier No. CGN476317
Chemglass CGN-4763-17 is a 1.2 mL cryogenic vial with a permanent laser-etched 2D barcode designed for reliable sample identification and handling. The vial is constructed from USP VI polypropylene and is sterilized by E-beam (SAL=10^-6). Packaged for laboratory use in multiple pack sizes to support inventory and storage workflows.
  • 1.2 mL capacity.
  • Laser-etched 2D barcode on the bottom for permanent identification.
  • USP VI polypropylene construction.
  • Sterile by E-beam; non-pyrogenic, DNase/RNase and human DNA free.
  • Self-standing round bottom with external thread.
  • Autoclavable.
  • Clear graduations and white marking area.
  • Special screw cap designed without a silicone ring to reduce contamination risk.
  • Packaged 50 per pack, 500 per box, 2000 per case.