Learn More
Chemglass Life Sciences Chemglass 2D Barcode 1.2 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile
SDP
Catalog No. 502733380
Encompass_Preferred
This 1.2 mL sterile cryogenic vial features a permanent laser 2D barcode on the bottom for reliable sample identification and rack-level scanning. The self-standing, round-bottom vial is made from USP VI polypropylene and includes a screw cap with a silicone O-ring to provide a secure seal. Vials are DNase/RNase and human DNA free, and are sterilized by E-beam (SAL = 10^-6).
  • 1.2 mL capacity.
  • Permanent laser 2D barcode on the vial bottom for rack-level scanning.
  • USP VI polypropylene construction.
  • Internal thread for secure cap fit.
  • Sterile; E-beam sterilized with SAL = 10^-6; non-pyrogenic.
  • DNase/RNase and human DNA free.
  • Screw cap with silicone O-ring for secure sealing.
  • Clear graduations and white marking area for sample labeling.
  • Self-standing, round-bottom design.
  • Autoclavable.
  • Packaged 9 x 9 per rack, 14 racks per case (1,134 vials per case).
Catalog No. 50-273-3380 Supplier Chemglass Life Sciences Supplier No. CGN476309
This 1.2 mL sterile cryogenic vial features a permanent laser 2D barcode on the bottom for reliable sample identification and rack-level scanning. The self-standing, round-bottom vial is made from USP VI polypropylene and includes a screw cap with a silicone O-ring to provide a secure seal. Vials are DNase/RNase and human DNA free, and are sterilized by E-beam (SAL = 10^-6).
  • 1.2 mL capacity.
  • Permanent laser 2D barcode on the vial bottom for rack-level scanning.
  • USP VI polypropylene construction.
  • Internal thread for secure cap fit.
  • Sterile; E-beam sterilized with SAL = 10^-6; non-pyrogenic.
  • DNase/RNase and human DNA free.
  • Screw cap with silicone O-ring for secure sealing.
  • Clear graduations and white marking area for sample labeling.
  • Self-standing, round-bottom design.
  • Autoclavable.
  • Packaged 9 x 9 per rack, 14 racks per case (1,134 vials per case).