Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 2.0 mL, Self-Standing, Internal Thread, Sterile, 2D Barcode, Racked
SDP
Catalog No. 502733378
Encompass_Preferred
The Chemglass CGN-4763-11 cryogenic vial is a 2.0 mL, sterile, self-standing polypropylene vial designed for secure long-term sample storage and high-throughput workflows. Each vial is permanently laser-marked with a unique 2D barcode for rapid rack-based scanning. The internal-thread screw cap with silicone O-ring provides a reliable seal; vials are E-beam sterilized (SAL 10^-6) and supplied racked for automation compatibility.
  • Permanent laser-etched 2D barcode on the vial bottom for fast, rack-based scanning.
  • Made from USP VI polypropylene, non-pyrogenic, and free of DNase/RNase and human DNA.
  • Clear graduations and white marking area for easy labeling and volume reading.
  • Autoclavable and E-beam sterilized (SAL 10^-6) for aseptic workflows.
  • Supplied racked (9 x 9 per rack), 12 racks per case (972 vials per case) for automation compatibility.
  • Self-standing round-bottom design for stable handling and storage.
Catalog No. 50-273-3378 Supplier Chemglass Life Sciences Supplier No. CGN476311
The Chemglass CGN-4763-11 cryogenic vial is a 2.0 mL, sterile, self-standing polypropylene vial designed for secure long-term sample storage and high-throughput workflows. Each vial is permanently laser-marked with a unique 2D barcode for rapid rack-based scanning. The internal-thread screw cap with silicone O-ring provides a reliable seal; vials are E-beam sterilized (SAL 10^-6) and supplied racked for automation compatibility.
  • Permanent laser-etched 2D barcode on the vial bottom for fast, rack-based scanning.
  • Made from USP VI polypropylene, non-pyrogenic, and free of DNase/RNase and human DNA.
  • Clear graduations and white marking area for easy labeling and volume reading.
  • Autoclavable and E-beam sterilized (SAL 10^-6) for aseptic workflows.
  • Supplied racked (9 x 9 per rack), 12 racks per case (972 vials per case) for automation compatibility.
  • Self-standing round-bottom design for stable handling and storage.