Learn More
Chemglass Life Sciences Chemglass 2.0 mL Cryogenic Vial, Self-Standing, External Thread, Sterile, Permanent 2D Barcode, Racked
SDP
Catalog No. 502733386
Encompass_Preferred
2D barcode 2.0 mL cryogenic vials provide reliable low-temperature sample storage and permanent identification. These self-standing, external-thread polypropylene vials are sterile and supplied in racked nests for organized handling and secure traceability.
  • 2.0 mL capacity for cryogenic storage
  • External thread for secure closure
  • Permanently marked 2D barcode on vial bottom for traceability
  • Sterile; E-beam sterilized, SAL=10-6
  • Made from USP VI polypropylene; autoclavable
  • Non-pyrogenic, DNase/RNase and human DNA free
  • Racked 9 x 9 per rack for easy handling; 12 racks per case
  • Solid one-piece construction to protect barcode at low temperatures
Catalog No. 50-273-3386 Supplier Chemglass Life Sciences Supplier No. CGN476303
2D barcode 2.0 mL cryogenic vials provide reliable low-temperature sample storage and permanent identification. These self-standing, external-thread polypropylene vials are sterile and supplied in racked nests for organized handling and secure traceability.
  • 2.0 mL capacity for cryogenic storage
  • External thread for secure closure
  • Permanently marked 2D barcode on vial bottom for traceability
  • Sterile; E-beam sterilized, SAL=10-6
  • Made from USP VI polypropylene; autoclavable
  • Non-pyrogenic, DNase/RNase and human DNA free
  • Racked 9 x 9 per rack for easy handling; 12 racks per case
  • Solid one-piece construction to protect barcode at low temperatures