Learn More
Chemglass Life Sciences Chemglass 2D Barcode 2.0 mL Cryogenic Vial, Self-Standing, External Thread, Sterile (50/pk)
SDP
Catalog No. 502733371
Encompass_Preferred
The Chemglass 2D Barcode 2.0 mL cryogenic vial provides secure, low-temperature sample storage with a permanent laser-etched 2D barcode for rapid traceability. Its self-standing, external-thread design and contamination-reducing screw cap make it suitable for biobanking and lab cryogenic workflows.
  • 2.0 mL capacity.
  • Laser-etched 2D barcode on the bottom for rapid scanning.
  • Self-standing vial with external thread.
  • Sterile: E-beam sterilized (SAL=10^-6); non-pyrogenic; DNase/RNase and human DNA free.
  • Made from USP VI polypropylene.
  • Packaging: 50 per pack, 500 per box, 2000 per case.
  • Autoclavable.
  • Clear graduations and white marking area for labeling.
  • Special screw cap designed without a silicone ring to reduce contamination risk.
Catalog No. 50-273-3371 Supplier Chemglass Life Sciences Supplier No. CGN476318
The Chemglass 2D Barcode 2.0 mL cryogenic vial provides secure, low-temperature sample storage with a permanent laser-etched 2D barcode for rapid traceability. Its self-standing, external-thread design and contamination-reducing screw cap make it suitable for biobanking and lab cryogenic workflows.
  • 2.0 mL capacity.
  • Laser-etched 2D barcode on the bottom for rapid scanning.
  • Self-standing vial with external thread.
  • Sterile: E-beam sterilized (SAL=10^-6); non-pyrogenic; DNase/RNase and human DNA free.
  • Made from USP VI polypropylene.
  • Packaging: 50 per pack, 500 per box, 2000 per case.
  • Autoclavable.
  • Clear graduations and white marking area for labeling.
  • Special screw cap designed without a silicone ring to reduce contamination risk.