Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 2.0 mL, Self-Standing, Internal Thread, Sterile, Laser 2D Barcode - Racked (10 x 10 per rack; 12 racks/case)
SDP
Catalog No. 502733377
Encompass_Preferred
Sterile 2.0 mL cryogenic vial with a permanent laser 2D barcode on the bottom for rapid rack scanning and inventory tracking. Self-standing, internal-thread design is supplied racked in nests (10 x 10 per rack, 12 racks per case) and is suitable for low-temperature storage and sample management.
  • 2.0 mL capacity.
  • Laser 2D barcode on vial bottom for scanning without removal from rack.
  • Self-standing, round-bottom design.
  • Internal thread with silicone O-ring cap for secure sealing.
  • USP VI polypropylene; non-pyrogenic and DNase/RNase free.
  • E-beam sterilized (SAL = 10^-6).
  • Autoclavable; clear graduations and white marking area.
Catalog No. 50-273-3377 Supplier Chemglass Life Sciences Supplier No. CGN476312
Sterile 2.0 mL cryogenic vial with a permanent laser 2D barcode on the bottom for rapid rack scanning and inventory tracking. Self-standing, internal-thread design is supplied racked in nests (10 x 10 per rack, 12 racks per case) and is suitable for low-temperature storage and sample management.
  • 2.0 mL capacity.
  • Laser 2D barcode on vial bottom for scanning without removal from rack.
  • Self-standing, round-bottom design.
  • Internal thread with silicone O-ring cap for secure sealing.
  • USP VI polypropylene; non-pyrogenic and DNase/RNase free.
  • E-beam sterilized (SAL = 10^-6).
  • Autoclavable; clear graduations and white marking area.