Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial - 2D Barcode, 3.0 mL, Self-Standing, External Thread, Sterile, Racked (9 x 9 per rack, 8 racks/case, 648 vials/case)
SDP
Catalog No. 502733385
Encompass_Preferred
Self-standing 3.0 mL cryogenic vials with a permanent laser 2D barcode on the bottom enable rapid, high-throughput sample identification while stored in racks. Vials are made from USP Class VI polypropylene, supplied sterile, and are suitable for low-temperature storage and autoclave sterilization. Supplied racked in 9 x 9 format for efficient handling.
  • Permanent laser 2D barcode on the bottom for rapid scanning in racks.
  • 3.0 mL capacity, self-standing design for easy handling and storage.
  • External thread with special screw cap designed without a silicone ring to reduce contamination risk.
  • Made from USP Class VI polypropylene; non-pyrogenic and DNase/RNase and human DNA free.
  • Sterile and autoclavable.
  • 100% barcode inspection to ensure unique and readable codes.
  • Clear graduations and white marking area for easy labeling.
  • 9 x 9 per rack, 8 racks per case (648 vials per case) for organized storage.
Catalog No. 50-273-3385 Supplier Chemglass Life Sciences Supplier No. CGN476304
Self-standing 3.0 mL cryogenic vials with a permanent laser 2D barcode on the bottom enable rapid, high-throughput sample identification while stored in racks. Vials are made from USP Class VI polypropylene, supplied sterile, and are suitable for low-temperature storage and autoclave sterilization. Supplied racked in 9 x 9 format for efficient handling.
  • Permanent laser 2D barcode on the bottom for rapid scanning in racks.
  • 3.0 mL capacity, self-standing design for easy handling and storage.
  • External thread with special screw cap designed without a silicone ring to reduce contamination risk.
  • Made from USP Class VI polypropylene; non-pyrogenic and DNase/RNase and human DNA free.
  • Sterile and autoclavable.
  • 100% barcode inspection to ensure unique and readable codes.
  • Clear graduations and white marking area for easy labeling.
  • 9 x 9 per rack, 8 racks per case (648 vials per case) for organized storage.