Learn More
Chemglass Life Sciences Chemglass 2D Barcode 4.0 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile - 9 x 9 rack; 648 vials/case
SDP
Catalog No. 502733375
Encompass_Preferred
These self-standing 4.0 mL cryogenic vials are manufactured from USP VI polypropylene and are permanently laser-marked with unique 2D barcodes for rapid, rack-based scanning. Vials are sterile, DNase/RNase and human DNA free, and include a silicone O-ring cap for secure sealing during low-temperature storage.
  • 4.0 mL volume.
  • USP VI polypropylene construction.
  • E-beam sterilized with SAL=10^-6; non-pyrogenic; DNase/RNase and human DNA free.
  • Internal thread with silicone O-ring screw cap.
  • Permanent laser 2D barcode on vial bottom; 100% inspected for uniqueness and readability.
  • 9 x 9 rack configuration; 8 racks per case (648 vials per case).
  • Round bottom, self-standing design with clear graduations and white marking area.
  • Autoclavable.
Catalog No. 50-273-3375 Supplier Chemglass Life Sciences Supplier No. CGN476314
These self-standing 4.0 mL cryogenic vials are manufactured from USP VI polypropylene and are permanently laser-marked with unique 2D barcodes for rapid, rack-based scanning. Vials are sterile, DNase/RNase and human DNA free, and include a silicone O-ring cap for secure sealing during low-temperature storage.
  • 4.0 mL volume.
  • USP VI polypropylene construction.
  • E-beam sterilized with SAL=10^-6; non-pyrogenic; DNase/RNase and human DNA free.
  • Internal thread with silicone O-ring screw cap.
  • Permanent laser 2D barcode on vial bottom; 100% inspected for uniqueness and readability.
  • 9 x 9 rack configuration; 8 racks per case (648 vials per case).
  • Round bottom, self-standing design with clear graduations and white marking area.
  • Autoclavable.