Learn More
Chemglass Life Sciences Chemglass Cryogenic vial, 5.0 mL, self-standing, external thread, sterile, laser-etched 2D barcode, racked (9 x 9), 486/case
SDP
Catalog No. 502733383
Encompass_Preferred
The Chemglass 5.0 mL cryogenic vial is a sterile, self-standing polypropylene vial designed for reliable long-term cryogenic storage and sample traceability. Each vial is permanently laser-etched with a unique 2D barcode on the bottom and is supplied racked for organized storage and compatibility with automated handling.
  • 5.0 mL capacity suitable for cryogenic storage.
  • Self-standing, round-bottom design for stable placement.
  • External thread with a special screw cap without silicone ring to reduce contamination risk.
  • Sterilized by E-beam with SAL=10^-6.
  • Made from USP VI polypropylene; autoclavable.
  • Laser-etched 2D barcode permanently marked on the vial bottom for unique identification.
  • Racked packaging (9 x 9 per rack), 6 racks per case, 486 vials per case.
  • Non-pyrogenic; DNase/RNase and human DNA free.
  • Clear graduations and white marking area for easy labeling.
Catalog No. 50-273-3383 Supplier Chemglass Life Sciences Supplier No. CGN476306
The Chemglass 5.0 mL cryogenic vial is a sterile, self-standing polypropylene vial designed for reliable long-term cryogenic storage and sample traceability. Each vial is permanently laser-etched with a unique 2D barcode on the bottom and is supplied racked for organized storage and compatibility with automated handling.
  • 5.0 mL capacity suitable for cryogenic storage.
  • Self-standing, round-bottom design for stable placement.
  • External thread with a special screw cap without silicone ring to reduce contamination risk.
  • Sterilized by E-beam with SAL=10^-6.
  • Made from USP VI polypropylene; autoclavable.
  • Laser-etched 2D barcode permanently marked on the vial bottom for unique identification.
  • Racked packaging (9 x 9 per rack), 6 racks per case, 486 vials per case.
  • Non-pyrogenic; DNase/RNase and human DNA free.
  • Clear graduations and white marking area for easy labeling.