Learn More
Chemglass Life Sciences 2D Barcode 5.0 mL Cryogenic Vial, Self-Standing, External Thread, Sterile, Bulk Packed (50/pk, 1,000/cs)
SDP
Catalog No. 502733369
Encompass_Preferred
Self-standing 5.0 mL cryogenic vial with a permanent laser 2D barcode on the bottom for reliable sample identification. Made from USP VI polypropylene, the vial features an external screw thread, clear graduations, and a silicone-free screw cap. Supplied sterile (E-beam sterilized, SAL=10^-6), non-pyrogenic, and DNase/RNase and human DNA free; packaged bulk (50 per pack, 1,000 per case).
  • Permanent laser 2D barcode for traceable sample tracking.
  • 5.0 mL capacity with clear graduations and white marking area.
  • External screw thread with silicone-free cap to reduce contamination risk.
  • Made from USP VI polypropylene for chemical compatibility.
  • Sterile, non-pyrogenic, and free of DNase, RNase, and human DNA.
  • Bulk packed format with 50 per pack and 1,000 per case for efficient stocking.
  • Round bottom, self-standing design suitable for cryogenic storage.
  • Autoclavable and inspected for barcode readability.
Catalog No. 50-273-3369 Supplier Chemglass Life Sciences Supplier No. CGN476321
Self-standing 5.0 mL cryogenic vial with a permanent laser 2D barcode on the bottom for reliable sample identification. Made from USP VI polypropylene, the vial features an external screw thread, clear graduations, and a silicone-free screw cap. Supplied sterile (E-beam sterilized, SAL=10^-6), non-pyrogenic, and DNase/RNase and human DNA free; packaged bulk (50 per pack, 1,000 per case).
  • Permanent laser 2D barcode for traceable sample tracking.
  • 5.0 mL capacity with clear graduations and white marking area.
  • External screw thread with silicone-free cap to reduce contamination risk.
  • Made from USP VI polypropylene for chemical compatibility.
  • Sterile, non-pyrogenic, and free of DNase, RNase, and human DNA.
  • Bulk packed format with 50 per pack and 1,000 per case for efficient stocking.
  • Round bottom, self-standing design suitable for cryogenic storage.
  • Autoclavable and inspected for barcode readability.