Learn More
Chemglass Life Sciences Chemglass 2D Barcode Cryogenic Vial, 5.0 mL, Self-Standing, Internal Thread, Sterile
SDP
Catalog No. 502733363
Encompass_Preferred
A sterile 5.0 mL cryogenic vial with an internal thread and self-standing design, featuring a permanent laser 2D barcode for fast, reliable sample identification and traceability.
  • 5.0 mL capacity.
  • Internal thread with self-standing base.
  • Permanent laser 2D barcode on the vial bottom for scan-ready identification.
  • Sterile; e-beam sterilized with SAL 10^-6.
  • USP VI polypropylene; non-pyrogenic, DNase/RNase and human DNA free.
  • Special screw cap without silicone ring to reduce contamination risk.
  • Clear graduations and white marking area for easy labeling.
  • Packaged 50 per pack, 250 per box, 1,000 per case.
Catalog No. 50-273-3363 Supplier Chemglass Life Sciences Supplier No. CGN476327
A sterile 5.0 mL cryogenic vial with an internal thread and self-standing design, featuring a permanent laser 2D barcode for fast, reliable sample identification and traceability.
  • 5.0 mL capacity.
  • Internal thread with self-standing base.
  • Permanent laser 2D barcode on the vial bottom for scan-ready identification.
  • Sterile; e-beam sterilized with SAL 10^-6.
  • USP VI polypropylene; non-pyrogenic, DNase/RNase and human DNA free.
  • Special screw cap without silicone ring to reduce contamination risk.
  • Clear graduations and white marking area for easy labeling.
  • Packaged 50 per pack, 250 per box, 1,000 per case.