Learn More
Description
The Barcode Reader for Thermo Scientific KingFisher Duo Prime is an optional accessory that allows users to track samples directly in the instrument’s internal software quickly and easily.
Specifications
Specifications
| Format | 1D and 2D |
| Description | 2D Barcode reader for KingFisher |
| For Use With (Equipment) | KingFisher Duo Prime |
| Quantity | Each |
Frequently Asked Questions (FAQs)
Yes, the KingFisher Duo Prime barcode reader is able to read the 2D barcode on the Matrix 500 µL ScrewTop Tubes. The barcode reader can generate an error if the tubes are in the tube rack and the barcode position is not able to be scanned. If this occurs, we recommend scanning each tube individually.
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.