Learn More
PixCell Medical™ HemoScreen 2D Barcode Scanner
Catalog No. 23123801
Change view
Click to view available options
Type:
Barcode Scanner
Catalog No. Type
23-123-801 Barcode Scanner
1 options
This item is not returnable. View return policy
Catalog No. 23-123-801 Supplier PixCell Medical™ Supplier No. ACS0000600
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
HemoScreen compatible 2D barcode reader used to scan blood sample ID and QC assay sheet recovery ranges.
  • Barcode scanner is recommended for scanning patient blood sample ID.
  • Sample ID can also be entered via user interface keyboard.
  • Barcode scanner highly recommended to upload QC assay sheet recovery ranges.
Specifications
Type Barcode Scanner