Learn More
Research Products International Corp 2D Platform Rocker with Mat, Ideal for Processing Western Blots
SDP
Catalog No. 501976993
Encompass
RPI Platform Mixers offer unique and outstanding features at an economical price. The 2D platform mixer provides the ideal rocking motion for processing Western Blots. Features include adjustable speed from an ultra-slow 2 rpm up to 30 rpm and an easily adjustable platform angle. The platform angle can be changed without using tools. Mixers feature electronic controls and maintenance free brushless motors for long term performance and dependability. Mixer can be used in a cold room or incubator at temperatures from +4° up to 65°.
Two year warranty.
Catalog No. 50-197-6993 Supplier Research Products International Corp Supplier No. 248553
RPI Platform Mixers offer unique and outstanding features at an economical price. The 2D platform mixer provides the ideal rocking motion for processing Western Blots. Features include adjustable speed from an ultra-slow 2 rpm up to 30 rpm and an easily adjustable platform angle. The platform angle can be changed without using tools. Mixers feature electronic controls and maintenance free brushless motors for long term performance and dependability. Mixer can be used in a cold room or incubator at temperatures from +4° up to 65°.
Two year warranty.