Learn More
JDM LAB SOLUTIONS LLC Digital 2D Rocker with Speed adjustment
Catalog No. NC2385873
Encompass
The 2D Platform Rocker is excellent for a gentle mix of solutions, reagents, blotting and staining applications. It has a high-quality motor for durability and ultra-quiet operation. Space saving design and simple to operate make this a popular item for any laboratory.FeaturesAll models come with a dimpled non-skid mat.Large platform and hooks for rubber strips accommodate many applications.Units provide high capacity with rack accessory.All units are compatible with cold room and incubator environment.NS-05A , NS-14UA models have speed adjustment, continuous and timer mode with automatic stop.NS-14UA model provide additional feature of tilt angle adjustment from 1-15°.
Catalog No. NC2385873 Supplier JDM LAB SOLUTIONS LLC Supplier No. NS05
The 2D Platform Rocker is excellent for a gentle mix of solutions, reagents, blotting and staining applications. It has a high-quality motor for durability and ultra-quiet operation. Space saving design and simple to operate make this a popular item for any laboratory.FeaturesAll models come with a dimpled non-skid mat.Large platform and hooks for rubber strips accommodate many applications.Units provide high capacity with rack accessory.All units are compatible with cold room and incubator environment.NS-05A , NS-14UA models have speed adjustment, continuous and timer mode with automatic stop.NS-14UA model provide additional feature of tilt angle adjustment from 1-15°.