Learn More
Alkali Scientific BenchRocker™ 2D Rocker, Variable Speed, 115V
SDP
Catalog No. 502335800
Encompass_Preferred
BenchRocker™ 2D Rocker offers adjustable speed and tilt for a wide range of applications, large 14" x 12" work surface dual platform accessory that doubles work surface, and is cold room and incubator safe.
Catalog No. 50-233-5800 Supplier Alkali Scientific Supplier No. RS7235
BenchRocker™ 2D Rocker offers adjustable speed and tilt for a wide range of applications, large 14" x 12" work surface dual platform accessory that doubles work surface, and is cold room and incubator safe.