Learn More
Cayman Chemical 2Deoxy2fluoroadenosIn5 2.5mg
SDP
Catalog No. 502919096
Encompass_Preferred
A fluorinated derivative of ATP; inhibits E. coli RNA polymerase (Ki = 200 µM)
Catalog No. 50-291-9096 Supplier Cayman Chemical Supplier No. 386672.5 MG
A fluorinated derivative of ATP; inhibits E. coli RNA polymerase (Ki = 200 µM)