Learn More
Cayman Chemical 2Deoxy3O Nmethylnthr 2.7mg
SDP
Catalog No. 502918992
Encompass_Preferred
A fluorescent nucleotide derivative; decreases inositol phosphate formation in CHO-K1 cells expressing the human purinergic P2Y12 receptor at 100 µM; has been used to study the interaction between cardiac troponin I and myofibrils; ex/em = 356/453 nm, respectively
Catalog No. 50-291-8992 Supplier Cayman Chemical Supplier No. 386702.7 MG
A fluorescent nucleotide derivative; decreases inositol phosphate formation in CHO-K1 cells expressing the human purinergic P2Y12 receptor at 100 µM; has been used to study the interaction between cardiac troponin I and myofibrils; ex/em = 356/453 nm, respectively