Learn More
Cayman Chemical 2DeoxyadenosIn5OdIphos 50mg
SDP
Catalog No. 502911902
Encompass_Preferred
A nucleotide diphosphate; inhibits the polymerization of ADP and CDP by E. coli or P. fluorescens polynucleotide phosphorylase at 0.23 mM; levels are increased in erythrocytes isolated from patients with adenosine deaminase deficiency and SCID
Catalog No. 50-291-1902 Supplier Cayman Chemical Supplier No. 3619550 MG
A nucleotide diphosphate; inhibits the polymerization of ADP and CDP by E. coli or P. fluorescens polynucleotide phosphorylase at 0.23 mM; levels are increased in erythrocytes isolated from patients with adenosine deaminase deficiency and SCID