Learn More
Cayman Chemical 2DeoxyadenosIn5OtrIpho 100mg
SDP
Catalog No. 502948438
Encompass_Preferred
A purine nucleotide; a substrate for DNA polymerase in the synthesis of DNA; a noncompetitive inhibitor of ribonucleotide reductases; has commonly been used in DNA synthesis in research applications; accumulates in adenosine deaminase deficiency
Catalog No. 50-294-8438 Supplier Cayman Chemical Supplier No. 32566100 MG
A purine nucleotide; a substrate for DNA polymerase in the synthesis of DNA; a noncompetitive inhibitor of ribonucleotide reductases; has commonly been used in DNA synthesis in research applications; accumulates in adenosine deaminase deficiency