Learn More
Cayman Chemical 2DeoxyadenosIn5OtrIpho 50mg
SDP
Catalog No. 502946397
Encompass_Preferred
A purine nucleotide; a substrate for DNA polymerase in the synthesis of DNA; a noncompetitive inhibitor of ribonucleotide reductases; has commonly been used in DNA synthesis in research applications; accumulates in adenosine deaminase deficiency
Catalog No. 50-294-6397 Supplier Cayman Chemical Supplier No. 3256650 MG
A purine nucleotide; a substrate for DNA polymerase in the synthesis of DNA; a noncompetitive inhibitor of ribonucleotide reductases; has commonly been used in DNA synthesis in research applications; accumulates in adenosine deaminase deficiency