Learn More
Cayman Chemical 2'-Deoxycytidine 5'-diphosphate (sodium salt hydrate), Purity: ≥95%, CAS: 151151-32-5, 10 mg
SDP
Catalog No. 502994964
Encompass_Preferred
A nucleotide diphosphate for various biochemical research applications including studies of DNA synthesis; substrate for nucleoside diphosphate kinases
Catalog No. 50-299-4964 Supplier Cayman Chemical Supplier No. 2298210 MG
A nucleotide diphosphate for various biochemical research applications including studies of DNA synthesis; substrate for nucleoside diphosphate kinases