Learn More
Cayman Chemical 2DeoxycytIdIn 5dIphospha 50mg
SDP
Catalog No. 502994996
Encompass_Preferred
A nucleotide diphosphate for various biochemical research applications including studies of DNA synthesis; substrate for nucleoside diphosphate kinases
Catalog No. 50-299-4996 Supplier Cayman Chemical Supplier No. 2298250 MG
A nucleotide diphosphate for various biochemical research applications including studies of DNA synthesis; substrate for nucleoside diphosphate kinases