Learn More
Cayman Chemical 2’-Deoxycytidine-5’-O-triphosphate (sodium salt), Purity: ≥95%, CAS: 109909-44-6, 100 mg
SDP
Catalog No. 502946837
Encompass_Preferred
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases
Catalog No. 50-294-6837 Supplier Cayman Chemical Supplier No. 33484100 MG
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases