Learn More
Cayman Chemical 2deoxyDGlucose6phosphate 100mg
SDP
Catalog No. 502965497
Encompass_Preferred
A derivative of 2-DG that is produced in mammalian cells by the action of hexokinase; accumulation of 2-DG6P in cells can inhibit glycolysis, leading to cell death
Catalog No. 50-296-5497 Supplier Cayman Chemical Supplier No. 17149100 MG
A derivative of 2-DG that is produced in mammalian cells by the action of hexokinase; accumulation of 2-DG6P in cells can inhibit glycolysis, leading to cell death