Learn More
Cayman Chemical 2-deoxy-D-Glucose-6-phosphate (sodium salt), Purity: ≥98%, 50 mg
SDP
Catalog No. 502967855
Encompass_Preferred
A derivative of 2-DG that is produced in mammalian cells by the action of hexokinase; accumulation of 2-DG6P in cells can inhibit glycolysis, leading to cell death
Catalog No. 50-296-7855 Supplier Cayman Chemical Supplier No. 1714950 MG
A derivative of 2-DG that is produced in mammalian cells by the action of hexokinase; accumulation of 2-DG6P in cells can inhibit glycolysis, leading to cell death