Learn More
Medchemexpress LLC (2e)-1-(3,4-dimethoxyphenyl)-5-fluoro-3-(4-fluorophenyl)-2-(2-fluorophenyl)pent-1-en-4-one bromide | 1390654-28-0 | >98.8% | 524.38 | 100 MG
SDP
Catalog No. 5000437182
Encompass_Preferred
KRN2 bromide is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM. It selectively suppresses the expression of pro-inflammatory genes, including Nos2 and Il6, without hindering high-salt-induced NFAT5 and its target gene expressions. KRN2 also dose-dependently inhibits NF-κB p65 binding to Nfat5 promoter 1 and directly blocks the interaction between NF-κB p65 and its DNA binding sequence in the upstream site of Nfat5 exon 1.
  • Effectively suppresses AIA in which innate immune cells play a predominant role.
  • Effectively suppresses CIA as well as AIA in mice.
  • Decreases the production of pro-inflammatory cytokines and autoantibodies.
  • Reduces macrophage infiltration.
Catalog No. 50-004-37182 Supplier Medchemexpress LLC Supplier No. HY112125A100MG
KRN2 bromide is a selective inhibitor of nuclear factor of activated T cells (NFAT5), with an IC50 of 0.1 μM. It selectively suppresses the expression of pro-inflammatory genes, including Nos2 and Il6, without hindering high-salt-induced NFAT5 and its target gene expressions. KRN2 also dose-dependently inhibits NF-κB p65 binding to Nfat5 promoter 1 and directly blocks the interaction between NF-κB p65 and its DNA binding sequence in the upstream site of Nfat5 exon 1.
  • Effectively suppresses AIA in which innate immune cells play a predominant role.
  • Effectively suppresses CIA as well as AIA in mice.
  • Decreases the production of pro-inflammatory cytokines and autoantibodies.
  • Reduces macrophage infiltration.