Learn More
Medchemexpress LLC (2E)-2-[1-(3-bromophenyl)ethylidene]α-(benzoylamino)-3,4-dihydro-4-oxo-1-phthalazineacidi... | 916170-19-9 | MFCD00327460 | 99.4% | C25H20BrN5O3 | 10MG
SDP
Catalog No. 5000261887
Encompass_Preferred
AC-55541 is a selective small-molecule agonist of protease-activated receptor 2 (PAR2) used as a research tool to probe PAR2-mediated signaling in vitro and in vivo. It activates PI hydrolysis and Ca2+ mobilization pathways and produces pronociceptive responses in animal models. Analytical documentation is provided and the compound is highly soluble in DMSO for assay preparation.
  • Selective protease-activated receptor 2 (PAR2) agonist.
  • Demonstrated activity in PI hydrolysis and Ca2+ mobilization assays (pEC50 ≈ 5.9-6.7).
  • Pronociceptive activity observed in vivo.
  • High purity (99.4%) suitable for biochemical assays.
  • Soluble in DMSO at ≥ 51 mg/mL for stock preparation.
  • COA and SDS available for quality and safety verification.
Catalog No. 50-002-61887 Supplier Medchemexpress LLC Supplier No. HY1435010MG
AC-55541 is a selective small-molecule agonist of protease-activated receptor 2 (PAR2) used as a research tool to probe PAR2-mediated signaling in vitro and in vivo. It activates PI hydrolysis and Ca2+ mobilization pathways and produces pronociceptive responses in animal models. Analytical documentation is provided and the compound is highly soluble in DMSO for assay preparation.
  • Selective protease-activated receptor 2 (PAR2) agonist.
  • Demonstrated activity in PI hydrolysis and Ca2+ mobilization assays (pEC50 ≈ 5.9-6.7).
  • Pronociceptive activity observed in vivo.
  • High purity (99.4%) suitable for biochemical assays.
  • Soluble in DMSO at ≥ 51 mg/mL for stock preparation.
  • COA and SDS available for quality and safety verification.