Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2E)-2-dodecenedioic acid | 6402-36-4 | 99.8% | 50 MG
SDP

Catalog No. 5000233013
Encompass_Preferred

Traumatic acid is a wound-healing phytohormone used as a research reagent to study collagen biosynthesis, wound repair signaling, apoptosis, and oxidative stress in cellular models. Supplied as a solid for laboratory research use only.

  • High purity (99.79%).
  • Molecular weight 228.28 g/mol.
  • Chemical formula C12H20O4.
  • Supplied as 50 MG of solid material for in vitro studies.
  • Suitable for studies of wound healing, collagen production, cancer cell viability, and oxidative stress pathways.

Catalog No. 50-002-33013 Supplier Medchemexpress LLC Supplier No. HY11935850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Traumatic acid is a wound-healing phytohormone used as a research reagent to study collagen biosynthesis, wound repair signaling, apoptosis, and oxidative stress in cellular models. Supplied as a solid for laboratory research use only.

  • High purity (99.79%).
  • Molecular weight 228.28 g/mol.
  • Chemical formula C12H20O4.
  • Supplied as 50 MG of solid material for in vitro studies.
  • Suitable for studies of wound healing, collagen production, cancer cell viability, and oxidative stress pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.