Learn More
TARGETMOL CHEMICALS INC 2E-3-1 3-Benzodioxol-5-yl-2-pr
SDP
Catalog No. 502877272
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (2E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic acid displays EC50 values of 73.5 uM against T. cruzi. trypomastigotes. Purity 99.72%
Catalog No. 50-287-7272 Supplier TARGETMOL CHEMICALS INC Supplier No. TN70745MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (2E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic acid displays EC50 values of 73.5 uM against T. cruzi. trypomastigotes. Purity 99.72%