Learn More
Medchemexpress LLC (2E)-3-[[4-(1,1-dimethylethyl)phenyl]sulfonyl]-2-propenenitrile | 196309-76-9 | MFCD01862602 | 100.0% | 249.33 g/mol | C13H15NO2S | 25 MG
SDP
Catalog No. 5000245844
Encompass_Preferred
BAY 11-7085 is a small-molecule inhibitor that blocks NF-κB activation by preventing phosphorylation and promoting stabilization of IκBα. It is used in cellular and biochemical research to probe NF-κB signaling, apoptosis, and ferroptosis pathways.
  • Inhibits NF-κB activation and phosphorylation of IκBα.
  • Stabilizes IκBα with a reported IC50 of 10 μM.
  • High reported purity (>99.9% by LCMS), suitable for research use.
  • Solid form with good solubility in DMSO and ethanol.
  • Commonly used in cellular and biochemical assays studying inflammatory and cell-death pathways.
Catalog No. 50-002-45844 Supplier Medchemexpress LLC Supplier No. HY1025725MG
BAY 11-7085 is a small-molecule inhibitor that blocks NF-κB activation by preventing phosphorylation and promoting stabilization of IκBα. It is used in cellular and biochemical research to probe NF-κB signaling, apoptosis, and ferroptosis pathways.
  • Inhibits NF-κB activation and phosphorylation of IκBα.
  • Stabilizes IκBα with a reported IC50 of 10 μM.
  • High reported purity (>99.9% by LCMS), suitable for research use.
  • Solid form with good solubility in DMSO and ethanol.
  • Commonly used in cellular and biochemical assays studying inflammatory and cell-death pathways.