Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2E,4E)-hexa-2,4-dien-1-ol | 17102-64-6 | MFCD00002925 | 98.8% | 98.14 g/mol | C6H10O | 1 G
SDP

Catalog No. 5000300645
Encompass_Preferred

(2E,4E)-Hexa-2,4-dien-1-ol is a small organic compound containing a conjugated diene and a primary alcohol. It is used as a biochemical assay reagent and reference standard in chemical research and flavor/aroma studies. Supplied for research use only.

  • High purity (98.81%).
  • Molecular formula C6H10O; molecular weight 98.14 g/mol.
  • DMSO soluble (200 mg/mL); ultrasonic recommended.
  • Appearance: colorless to light yellow; solid below ~28°C, liquid above ~33°C.
  • Long-term storage stability: pure form -20°C for up to 3 years.
  • COA and SDS available for quality and safety information.

Catalog No. 50-003-00645 Supplier Medchemexpress LLC Supplier No. HYW0176901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2E,4E)-Hexa-2,4-dien-1-ol is a small organic compound containing a conjugated diene and a primary alcohol. It is used as a biochemical assay reagent and reference standard in chemical research and flavor/aroma studies. Supplied for research use only.

  • High purity (98.81%).
  • Molecular formula C6H10O; molecular weight 98.14 g/mol.
  • DMSO soluble (200 mg/mL); ultrasonic recommended.
  • Appearance: colorless to light yellow; solid below ~28°C, liquid above ~33°C.
  • Long-term storage stability: pure form -20°C for up to 3 years.
  • COA and SDS available for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.