Learn More
Cayman Chemical 2-Ethyl-5,7-dimethyl-3H-imidazo[4,5-b]pyridine, Purity: ≥98%, CAS: 133240-06-9, 100 mg
SDP
Catalog No. 502965503
Encompass_Preferred
A nonpeptidic angiotensin II receptor antagonist whose imidazo[4,5-b]pyridine scaffold has been used to examine a variety of positional substitutions in the development of orally active, long duration antihypertensive agents
Catalog No. 50-296-5503 Supplier Cayman Chemical Supplier No. 17645100 MG
A nonpeptidic angiotensin II receptor antagonist whose imidazo[4,5-b]pyridine scaffold has been used to examine a variety of positional substitutions in the development of orally active, long duration antihypertensive agents