Learn More
AdipoGen 2F-2 4-dihydroxybenzaldehyde
SDP
Supplier:  AdipoGen CDXD0330M250
Encompass_Preferred
Chemical. CAS 209541-27-5. Formula C7H4F2O3. MW 174.1. Synthetic. Building block for synthesis. Used for the preparation of fluorescent probes.
Catalog No. 50-256-1116