Learn More
Cayman Chemical 2F-Peracetyl-Fucose 100mg
SDP
Catalog No. 502990243
Encompass_Preferred
A protein fucosylation inhibitor; inhibits fucosylation of IgG1 monoclonal antibodies produced by CHO-DG44 cells at 50 µM; reduces intracellular GDP-fucose levels in CHO-DG44 cells; decreases intracellular fucose levels in A. thaliana seedlings at 100 µM; decreases root length in A. thaliana seedlings
Catalog No. 50-299-0243 Supplier Cayman Chemical Supplier No. 30865100 MG
Add to Cart
Add to Cart
A protein fucosylation inhibitor; inhibits fucosylation of IgG1 monoclonal antibodies produced by CHO-DG44 cells at 50 µM; reduces intracellular GDP-fucose levels in CHO-DG44 cells; decreases intracellular fucose levels in A. thaliana seedlings at 100 µM; decreases root length in A. thaliana seedlings