Learn More
Medchemexpress LLC 2F-Peracetyl-Fucose 10mM 1mL
SDP
Catalog No. 5000205559
Encompass_Preferred
2F-Peracetyl-Fucose (1 3 4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranos) acts as a potent fucosyltransferase (FUT) inhibitor[1]
Catalog No. 50-002-05559 Supplier Medchemexpress LLC Supplier No. HYW09660010MM/1ML
2F-Peracetyl-Fucose (1 3 4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranos) acts as a potent fucosyltransferase (FUT) inhibitor[1]