Learn More
Medchemexpress LLC 2F-Peracetyl-Fucose 25mg
SDP
Catalog No. 5000204562
Encompass_Preferred
2F-Peracetyl-Fucose (1 3 4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranos) acts as a potent fucosyltransferase (FUT) inhibitor[1]
Catalog No. 50-002-04562 Supplier Medchemexpress LLC Supplier No. 25MGHYW096600
2F-Peracetyl-Fucose (1 3 4-Tri-O-acetyl-2-deoxy-2-fluoro-L-fucopyranos) acts as a potent fucosyltransferase (FUT) inhibitor[1]