Learn More
Cayman Chemical 2fluoro PalmITC ACD mthyl 10mg
SDP
Catalog No. 502934858
Encompass_Preferred
A monofluorinated form of palmitic acid methyl ester; intermediate in the synthesis of 2-iodoaldehydes, monofluorinated fatty acids, and polyfluoro ketones
Catalog No. 50-293-4858 Supplier Cayman Chemical Supplier No. 2452610 MG
A monofluorinated form of palmitic acid methyl ester; intermediate in the synthesis of 2-iodoaldehydes, monofluorinated fatty acids, and polyfluoro ketones