Learn More
Cayman Chemical 2-Fluoro-4-iodo benzonitrile, Purity: ≥98%, CAS: 137553-42-5, 500 mg
SDP
Catalog No. 502992226
Encompass_Preferred
A building block; has been used in the synthesis of Li-TryR dimerization and oxidoreductase activity inhibitors; has been used in the synthesis of TRPA1 antagonists
Catalog No. 50-299-2226 Supplier Cayman Chemical Supplier No. 30483500 MG
A building block; has been used in the synthesis of Li-TryR dimerization and oxidoreductase activity inhibitors; has been used in the synthesis of TRPA1 antagonists