Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-1,2-oxazine derivative (TFA salt) | 96.0% | 1165.34 g/mol | C57H87F3N8O14 | 25 MG
SDP

Catalog No. 5000203420
Encompass_Preferred

NHS-modified monomethyl auristatin F (TFA) is an NHS-activated MMAF derivative supplied as a trifluoroacetate salt and used as an intermediate in antibody-drug conjugate (ADC) linker synthesis. It is provided for research applications and is intended for building drug-linker conjugates.

  • NHS-activated derivative suitable for conjugation to amines.
  • Intended as an intermediate for antibody-drug conjugate synthesis.
  • Purity around 96.0% by HPLC.
  • Molecular weight approximately 1165.34 g/mol.
  • Provided as a TFA salt with formula C57H87F3N8O14.

Catalog No. 50-002-03420 Supplier Medchemexpress LLC Supplier No. HY139325A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHS-modified monomethyl auristatin F (TFA) is an NHS-activated MMAF derivative supplied as a trifluoroacetate salt and used as an intermediate in antibody-drug conjugate (ADC) linker synthesis. It is provided for research applications and is intended for building drug-linker conjugates.

  • NHS-activated derivative suitable for conjugation to amines.
  • Intended as an intermediate for antibody-drug conjugate synthesis.
  • Purity around 96.0% by HPLC.
  • Molecular weight approximately 1165.34 g/mol.
  • Provided as a TFA salt with formula C57H87F3N8O14.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.