Learn More
Medchemexpress LLC 2H-1-Benzazepin-2-one, 6-fluoro-1,3,4,5-tetrahydro-3-[4-[3-methoxy-4-(4-methyl-1H-imidazol-1-yl)phenyl]-1H-1,2,3-triazol-1-yl]-1-(2,2,2-trifluoroethyl)- | 1093978-89-2 | >98.8% | C25H22F4N6O2 | 50 MG
SDP
Catalog No. 5000359294
Encompass_Preferred
This is a potent and selective modulator for the study of Alzheimer's disease. It has been shown to inhibit gamma-secretase activity in various cell lines, leading to a decrease in amyloid beta levels.
  • Potent and selective modulation of gamma-secretase.
  • Suitable for research into Alzheimer's disease.
  • Demonstrates cellular effects in HEK293, HeLa, and SH-SY5Y cell lines.
  • High purity of 98.80%.
  • Available in various quantities, including free samples.
  • Detailed documentation including data sheet, SDS, and handling instructions.
  • Soluble in DMSO for in vitro applications.
  • In vivo dissolution protocols provided.
Catalog No. 50-003-59294 Supplier Medchemexpress LLC Supplier No. HY5075450MG
This is a potent and selective modulator for the study of Alzheimer's disease. It has been shown to inhibit gamma-secretase activity in various cell lines, leading to a decrease in amyloid beta levels.
  • Potent and selective modulation of gamma-secretase.
  • Suitable for research into Alzheimer's disease.
  • Demonstrates cellular effects in HEK293, HeLa, and SH-SY5Y cell lines.
  • High purity of 98.80%.
  • Available in various quantities, including free samples.
  • Detailed documentation including data sheet, SDS, and handling instructions.
  • Soluble in DMSO for in vitro applications.
  • In vivo dissolution protocols provided.