Learn More
Medchemexpress LLC 2H-1-Benzopyran-2-one-5,6,7,8-d4 | 185056-83-1 | 150.17 | 25 MG
SDP
Catalog No. 5000389275
Encompass_Preferred
Coumarin-d4 is a deuterium-labeled form of Coumarin, which is the primary bioactive ingredient found in Radix Glehniae. It exhibits various pharmacological activities, including anticancer, anti-inflammatory, and antiviral properties.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-89275 Supplier Medchemexpress LLC Supplier No. 25MGHYN0709S
Coumarin-d4 is a deuterium-labeled form of Coumarin, which is the primary bioactive ingredient found in Radix Glehniae. It exhibits various pharmacological activities, including anticancer, anti-inflammatory, and antiviral properties.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS