Learn More
Medchemexpress LLC 2H-1-Benzopyran-2-one, 6-bromo- | 19063-55-9 | 225.04 | 10 G
SDP
Catalog No. 5000446049
Encompass_Preferred
6-Bromocoumarin (6-Bromochromen-2-one) is a chemical compound identified as a potential anti-bacterial agent. It has been noted for not inhibiting 17β-HSD1 and for showing affinity for α and β estrogen receptors.
  • Potential anti-bacterial agent
  • Does not inhibit 17β-HSD1
  • Shows affinity for α and β estrogen receptors
Catalog No. 50-004-46049 Supplier Medchemexpress LLC Supplier No. HYW00518610G
6-Bromocoumarin (6-Bromochromen-2-one) is a chemical compound identified as a potential anti-bacterial agent. It has been noted for not inhibiting 17β-HSD1 and for showing affinity for α and β estrogen receptors.
  • Potential anti-bacterial agent
  • Does not inhibit 17β-HSD1
  • Shows affinity for α and β estrogen receptors