Learn More
Medchemexpress LLC 2H-1-Benzopyran-2-one, 7-[[4-amino-6-[(2-methylphenyl)amino]-1,3,5-triazin-2-yl]methoxy]-4-methyl-. | 874755-26-7 | 98.3% | 389.41 g·mol⁻¹ | C21H19N5O3 | 5 MG
SDP
Catalog No. 5000263510
Encompass_Preferred
GPR40/FFAR1 modulator 1 is a small-molecule agonist and allosteric modulator of the Gq-coupled free fatty acid receptor 1 (GPR40/FFAR1). It is supplied for research use to probe receptor pharmacology, ligand modulation, and downstream signaling in cellular assays. Supplied as an off-white to light-yellow solid with characterized purity and molecular properties.
  • Acts as an agonist and allosteric modulator of GPR40/FFAR1.
  • Suitable for receptor pharmacology and signaling studies.
  • Provided as a solid and as a 10 mM solution in DMSO.
  • High purity (≈98.3%) for consistent experimental results.
  • Molecular formula C21H19N5O3 and molecular weight 389.41 g·mol⁻¹.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-63510 Supplier Medchemexpress LLC Supplier No. HY1117635MG
GPR40/FFAR1 modulator 1 is a small-molecule agonist and allosteric modulator of the Gq-coupled free fatty acid receptor 1 (GPR40/FFAR1). It is supplied for research use to probe receptor pharmacology, ligand modulation, and downstream signaling in cellular assays. Supplied as an off-white to light-yellow solid with characterized purity and molecular properties.
  • Acts as an agonist and allosteric modulator of GPR40/FFAR1.
  • Suitable for receptor pharmacology and signaling studies.
  • Provided as a solid and as a 10 mM solution in DMSO.
  • High purity (≈98.3%) for consistent experimental results.
  • Molecular formula C21H19N5O3 and molecular weight 389.41 g·mol⁻¹.
  • Intended for research use only; not for human or veterinary use.