Learn More
Medchemexpress LLC 2H-1-benzopyran-2-one, 7-(beta-D-glucopyranosyloxy)-4-methyl- | 18997-57-4 | MFCD00063694 | 99.9% | 338.31 g/mol | C16H18O8 | 10MM 1ML
SDP
Catalog No. 5000237691
Encompass_Preferred
4-Methylumbelliferyl β-D-glucopyranoside is a coumarin-based fluorogenic substrate used to measure β-glucosidase and related enzyme activities. Enzymatic hydrolysis releases the fluorescent 4-methylumbelliferone (4-MU), which emits near 445-454 nm; excitation is pH-dependent. The product is supplied as a 10 mM solution in DMSO (1 mL) and is suitable for fluorescence-based biochemical assays.
Catalog No. 50-002-37691 Supplier Medchemexpress LLC Supplier No. HY12363310MM 1ML
4-Methylumbelliferyl β-D-glucopyranoside is a coumarin-based fluorogenic substrate used to measure β-glucosidase and related enzyme activities. Enzymatic hydrolysis releases the fluorescent 4-methylumbelliferone (4-MU), which emits near 445-454 nm; excitation is pH-dependent. The product is supplied as a 10 mM solution in DMSO (1 mL) and is suitable for fluorescence-based biochemical assays.