Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2h-1-benzopyran-2-one, 7-ethoxy-4-methyl- | 87-05-8 | MFCD00016971 | 99.4% | 25 G
SDP

Catalog No. 5000414206
Encompass_Preferred

7-Ethoxy-4-methylcoumarin is a coumarin derivative that serves as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6. It is intended for research use only and not for sale to patients.

  • Coumarin derivative.
  • Functions as a substrate probe for specific cytochrome P450 enzymes.
  • Useful in research and analytical applications.
  • Targets CYP1A1, CYP2B4, and CYP2B6.
  • Can be classified as a fluorescent dye.
  • Used in labeling chemistry.
  • A metabolic enzyme/protease.

Catalog No. 50-004-14206 Supplier Medchemexpress LLC Supplier No. 25GHYD0092
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ethoxy-4-methylcoumarin is a coumarin derivative that serves as a substrate probe for mammalian cytochromes P450 1A1, 2B4, and 2B6. It is intended for research use only and not for sale to patients.

  • Coumarin derivative.
  • Functions as a substrate probe for specific cytochrome P450 enzymes.
  • Useful in research and analytical applications.
  • Targets CYP1A1, CYP2B4, and CYP2B6.
  • Can be classified as a fluorescent dye.
  • Used in labeling chemistry.
  • A metabolic enzyme/protease.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.